2,3,4-Trimethoxyphenylboronic acid

Leverantör: Apollo Scientific
Varning

Synonyms: (2,3,4-Trimethoxyphenyl)metaboric acid, 2,3,4-Trimethoxybenzeneboronic acid, Dihydroxy(2,3,4-trimethoxyphenyl)borane, (2,3,4-Trimethoxyphenyl)boronic acid

OR10593-1G OR10593-5G
APOSOR10593-1GEA 620 SEK
APOSOR10593-1G APOSOR10593-5G
2,3,4-Trimethoxyphenylboronic acid
2,3,4-Trimethoxyphenylboronic acid
Formel: C₉H₁₃BO₅
MW: 212,01 g/mol
Smältpunkt: 74…79 °C
Storage Temperature: Kylskåp
MDL Number: MFCD01318183
CAS nummer: 118062-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR