2-Phenoxyphenylboronic acid 98%
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
2-Phenoxybenzeneboronic acid
OR10577-10G
OR10577-5G
APOSOR10577-10GEA
1300
SEK
APOSOR10577-10G
APOSOR10577-5G
2-Phenoxyphenylboronic acid 98%
2-Phenoxyphenylboronic acid
Formel:
C₁₂H₁₁BO₃ MW: 214,03 g/mol Smältpunkt: 96…100 °C Storage Temperature: Kylskåp |
MDL Number:
MFCD01001592 CAS nummer: 108238-09-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...