2-Methyl-5-nitrophenylboronic acid 96%

Leverantör: Apollo Scientific
Varning

Synonyms: 2-Methyl-5-nitrobenzeneboronic acid

OR10548-1G OR10548-5G
APOSOR10548-1GEA 1050 SEK
APOSOR10548-1G APOSOR10548-5G
2-Methyl-5-nitrophenylboronic acid 96%
2-Methyl-5-nitrophenylboronic acid
Formel: C₇H₈BNO₄
MW: 180,96 g/mol
Smältpunkt: 60…66 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00757435
CAS nummer: 100960-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR