1-Methylimidazole-5-carboxylic acid
Leverantör: Apollo Scientific
Varning
Synonyms: 1-Methylimidazole-5-carboxylic Acid
OR10543-1G OR10543-25G OR10543-5G
APOSOR10543-1GEA 175 SEK
APOSOR10543-1G APOSOR10543-25G APOSOR10543-5G
1-Methylimidazole-5-carboxylic acid
1-Methylimidazole-5-carboxylic acid
Formel: C₅H₆N₂O₂
MW: 126,1133 g/mol
Smältpunkt: 247…250 °C (dec.)
Storage Temperature: Kylskåp
MDL Number: MFCD00955677
CAS nummer: 41806-40-0