4-Isobutylphenylboronic acid 98%
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
4-Isobutylbenzeneboronic acid
OR10519-1G
OR10519-5G
APOSOR10519-1GEA
175
SEK
APOSOR10519-1G
APOSOR10519-5G
4-Isobutylphenylboronic acid 98%
4-Isobutylphenylboronic acid
Formel:
C₁₀H₁₅BO₂ MW: 178,04 g/mol Smältpunkt: 117…120 °C Storage Temperature: Kylskåp |
MDL Number:
MFCD01632198 CAS nummer: 153624-38-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...