2-Iodo-1-methylimidazole
Leverantör: Apollo Scientific
Varning
Synonyms: 2-Iodo-1-methyl-1H-imidazole
OR10510-1G OR10510-5G OR10510-10G
APOSOR10510-1GEA 855 SEK
APOSOR10510-1G APOSOR10510-5G APOSOR10510-10G
2-Iodo-1-methylimidazole
2-Iodo-1-methylimidazole
Formel: C₄H₅IN₂
MW: 208 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00661606
CAS nummer: 37067-95-1