2-(Ethoxycarbonyl)phenylboronic acid 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 2-(Ethoxycarbonyl)benzeneboronic acid, (2-(Ethoxycarbonyl)phenyl)boronic acid
OR10468-25G OR10468-5G
APOSOR10468-25GEA 1250 SEK
APOSOR10468-25G APOSOR10468-5G
2-(Ethoxycarbonyl)phenylboronic acid 97%
2-(Ethoxycarbonyl)phenylboronic acid
Formel: C₉H₁₁BO₄
MW: 193,99 g/mol
Smältpunkt: 128…136 °C
Storage Temperature: Kylskåp
MDL Number: MFCD02179453
CAS nummer: 380430-53-5