5-Chloro-2-methylphenylboronic acid 97%
Leverantör: Apollo Scientific
Varning
Synonyms: (5-Chloro-2-methylphenyl)boronic acid, 5-Chloro-2-methylbenzeneboronic acid
OR10415-1G OR10415-5G OR10415-10G
APOSOR10415-1GEA 175 SEK
APOSOR10415-1G APOSOR10415-5G APOSOR10415-10G
5-Chloro-2-methylphenylboronic acid 97%
5-Chloro-2-methylphenylboronic acid
Formel: C₇H₈BClO₂
MW: 170,40 g/mol
Smältpunkt: 184…186 °C
Storage Temperature: Omgivande
MDL Number: MFCD03411939
CAS nummer: 148839-33-2