3-Carboxy-5-nitrophenylboronic acid

Leverantör: Apollo Scientific
Varning

Synonyms: 3-Carboxy-5-nitrobenzeneboronic acid, 3-Borono-5-nitrobenzoic acid

OR10400-10G OR10400-1G OR10400-5G
APOSOR10400-10GEA 875 SEK
APOSOR10400-10G APOSOR10400-1G APOSOR10400-5G
3-Carboxy-5-nitrophenylboronic acid
3-Carboxy-5-nitrophenylboronic acid
Formel: C₇H₆BNO₆
MW: 210,94 g/mol
Smältpunkt: 229…232 °C
Storage Temperature: Omgivande
MDL Number: MFCD00757433
CAS nummer: 101084-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR