2-Amino-5-nitrothiazole 97%

Leverantör: Apollo Scientific
Varning

Synonyms: 5-Nitrothiazol-2-ylamine, 5-Nitrothiazol-2-amine

OR10323-100G OR10323-1KG OR10323-500G
APOSOR10323-100GEA 175 SEK
APOSOR10323-100G APOSOR10323-1KG APOSOR10323-500G
2-Amino-5-nitrothiazole 97%
2-Amino-5-nitrothiazole
Formel: C₃H₃N₃O₂S
MW: 145,14 g/mol
Smältpunkt: 195…202 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00005326
CAS nummer: 121-66-4
EINECS: 204-490-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR