Methyl Indole-2-carboxylate 97%

Leverantör: Apollo Scientific
Varning

Synonyms: Methyl 1H-indole-2-carboxylate, Indole-2-carboxylic Acid Methyl Ester

OR10191-100G OR10191-25G
APOSOR10191-100GEA 1800 SEK
APOSOR10191-100G APOSOR10191-25G
Methyl Indole-2-carboxylate 97%
Methyl Indole-2-carboxylate
Formel: C₁₀H₉NO₂
MW: 175,19 g/mol
Smältpunkt: 151…153 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00460779
CAS nummer: 1202-04-6
EINECS: 214-861-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR