4-Nitrothiophenol 95%
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
4-Nitrobenzene-1-thiol, 4-Nitrobenzenethiol, 1-Mercapto-4-nitrobenzene, p-Nitrothiophenol
OR0779-25G
OR0779-50G
APOSOR0779-25GEA
2850
SEK
APOSOR0779-25G
APOSOR0779-50G
4-Nitrothiophenol 95%
4-Nitrothiophenol
Formel:
C₆H₅NO₂S MW: 155,18 g/mol Smältpunkt: 72…77 °C Storage Temperature: Kylskåp |
MDL Number:
MFCD00007343 CAS nummer: 1849-36-1 EINECS: 217-436-4 UN: 3335 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...