D-(+)-Proline
Leverantör: Apollo Scientific
Varning
Synonyms: H-D-Pro-OH, (R)-(+)-Proline, D-Proline
OR0718-100G OR0718-25G OR0718-5G
APOSOR0718-100GEA 1825 SEK
APOSOR0718-100G APOSOR0718-25G APOSOR0718-5G
D-(+)-Proline
D-(+)-Proline
Chiral intermediate for pharmaceuticals and agrochemicals.
Formel: C₅H₉NO₂
MW: 115,13 g/mol
Smältpunkt: 223 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00064317
CAS nummer: 344-25-2
EINECS: 206-452-7