5-(Phenylethynyl)-2-furoic acid, Technical Grade
Leverantör: Apollo Scientific
Varning
OR0707-250MG OR0707-1G
APOSOR0707-250MGEA 685 SEK
APOSOR0707-250MG APOSOR0707-1G
5-(Phenylethynyl)-2-furoic acid, Technical Grade
5-(Phenylethynyl)-2-furoic acid
Formel: C₁₃H₈O₃
Smältpunkt: 212…213 °C
Storage Temperature: Omgivande
MDL Number: MFCD00541949
CAS nummer: 130423-83-5