2,4-Dichlorophenyl isocyanate
Leverantör: Apollo Scientific
Fara
Synonyms: 2,4-Dichlorophenylisocyantae
OR0363-25G OR0363-5G
APOSOR0363-25GEA 1600 SEK
APOSOR0363-25G APOSOR0363-5G
2,4-Dichlorophenyl isocyanate
2,4-Dichlorophenyl isocyanate
  • May form small amount of sediment.
Formel: C₇H₃Cl₂NO
MW: 188,01 g/mol
Kokpunkt: 80 °C/1mmHg
Smältpunkt: 57…62 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002001
CAS nummer: 2612-57-9
EINECS: 220-040-4
UN: 2250
ADR: 6.1,II