2,4-Dichlorobenzoyl chloride

Leverantör: Apollo Scientific
Fara

Synonyms: 2,4-Dichlorobenzene-1-carbonyl chloride

OR0342-250G OR0342-50G
APOSOR0342-250GEA 1325 SEK
APOSOR0342-250G APOSOR0342-50G
2,4-Dichlorobenzoyl chloride
2,4-Dichlorobenzoyl chloride
Formel: C₇H₃Cl₃O
MW: 209,46 g/mol
Kokpunkt: 150 °C/34mmHg
Smältpunkt: 16…20 °C
Densitet: 1,494 g/cm³ (25 °C)
Flampunkt: 137 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000661
CAS nummer: 89-75-8
EINECS: 201-938-4
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR