ω-Bromo-3-cyanoacetophenone 98%
Leverantör: Apollo Scientific
|
Fara
|
Synonyms:
3-(2-Bromoacetyl)benzonitrile, 2-Bromo-3'-cyanoacetophenone, 3-(Bromoacetyl)benzonitrile, 3-Cyanophenacyl bromide, Bromomethyl m-cyanophenyl ketone, α-Bromo-3-cyanoacetophenone, 3-(2-Bromoacetyl)benzenecarbonitrile
OR0311-25G
OR0311-5G
APOSOR0311-25GEA
4275
SEK
APOSOR0311-25G
APOSOR0311-5G
ω-Bromo-3-cyanoacetophenone 98%
ω-Bromo-3-cyanoacetophenone
Formel:
C₉H₆BrNO MW: 224,06 g/mol Smältpunkt: 65…67 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00833267 CAS nummer: 50916-55-7 UN: 2923 ADR: 8+6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...