trans-4-(4-Chlorophenyl)cyclohexane-1-carboxylic acid 98%
Leverantör: Apollo Scientific
Varning
Synonyms: trans-4-(4-Chlorophenyl)cyclohexanecarboxylic acid
OR0279-5G OR0279-1G OR0279-25G
APOSOR0279-5GEA 670 SEK
APOSOR0279-5G APOSOR0279-1G APOSOR0279-25G
trans-4-(4-Chlorophenyl)cyclohexane-1-carboxylic acid 98%
trans-4-(4-Chlorophenyl)cyclohexane-1-carboxylic acid
Formel: C₁₃H₁₅ClO₂
MW: 238,71 g/mol
Smältpunkt: 249…255 °C
Storage Temperature: Omgivande
MDL Number: MFCD06797626
CAS nummer: 49708-81-8