6-Chloro-o-cresol 98%
Leverantör: Apollo Scientific
|
Fara
|
Synonyms:
1-Chloro-2-hydroxy-3-methylbenzene, 2-Chloro-6-methylphenol
OR0259-25G
OR0259-5G
APOSOR0259-25GEA
4675
SEK
APOSOR0259-25G
APOSOR0259-5G
6-Chloro-o-cresol 98%
6-Chloro-o-cresol
Formel:
C₇H₇ClO MW: 142,58 g/mol Kokpunkt: 75…76 °C/12mmHg Densitet: 1,201 g/cm³ (25 °C) Flampunkt: 72 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00020000 CAS nummer: 87-64-9 EINECS: 201-760-8 UN: 2669 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...