2-Allylbenzonitrile

Leverantör: Apollo Scientific
Varning

OR01935-100MG OR01935-500MG
APOSOR01935-100MGEA 2150 SEK
APOSOR01935-100MG APOSOR01935-500MG
2-Allylbenzonitrile
2-Allylbenzonitrile
Formel: C₁₀H₉N
Storage Temperature: Omgivande
CAS nummer: 61463-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR