2,6-Dihydroxy-3-nitrobenzonitrile

Leverantör: Apollo Scientific

OR017404-1G
APOSOR017404-1GEA 1475 SEK
APOSOR017404-1G
2,6-Dihydroxy-3-nitrobenzonitrile
2,6-Dihydroxy-3-nitrobenzonitrile
Formel: C₇H₄N₂O₄
Smältpunkt: 238 °C
Storage Temperature: Omgivande
CAS nummer: 233585-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR