1-(2-Pyridyl)ethylamine
Leverantör: Apollo Scientific
Fara
Synonyms: 2-(1-Aminoethyl)pyridine
OR01560-1G OR01560-5G OR01560-25G
APOSOR01560-1GEA 360 SEK
APOSOR01560-1G APOSOR01560-5G APOSOR01560-25G
1-(2-Pyridyl)ethylamine
1-(2-Pyridyl)ethylamine
Formel: C₇H₁₀N₂
MW: 122,17 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD05215238
CAS nummer: 42088-91-5
UN: 2735
ADR: 8,III