Nɑ-Benzoyl-L-arginine ethyl ester hydrochloride
Leverantör: Apollo Scientific
Synonyms: Bz-Arg-OEt.HCl, Nα-Benzoyl-L-arginine Ethyl Ester HCl
OR0094-100G OR0094-25G OR0094-10G
APOSOR0094-100GEA 2925 SEK
APOSOR0094-100G APOSOR0094-25G APOSOR0094-10G
Nɑ-Benzoyl-L-arginine ethyl ester hydrochloride
Nalpha-Benzoyl-L-arginine ethyl ester hydrochloride
Formel: C₁₅H₂₂N₄O₃
MW: 342.82 g/mol
Smältpunkt: 128…130 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00012579
CAS nummer: 2645-08-1