Yttrium(III)hexahydrat 99,9%
Leverantör: Apollo Scientific
Varning
Synonyms: Yttrium trichloride hexahydrate
IN3841-100G IN3841-25G
APOSIN3841-100GEA 3175 SEK
APOSIN3841-100G APOSIN3841-25G
Yttrium(III)hexahydrat 99,9%
Yttrium(III)hexahydrat
Formel: YCl₃·6H₂O
MW: 303,36 g/mol
Smältpunkt: 100 °C (dec.)
Densitet: 2,18 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00149941
CAS nummer: 10025-94-2