Ruthenium(IV) oxide 99.5+%
Leverantör: Apollo Scientific
Fara
Synonyms: Ruthenium dioxide, Ruthenium tetroxide
IN3064-1G IN3064-5G
APOSIN3064-1GEA 1475 SEK
APOSIN3064-1G APOSIN3064-5G
Ruthenium(IV) oxide 99.5+%
Ruthenium(IV) oxide
Formel: O₂Ru
Smältpunkt: 1200 °C (subl.)
Densitet: 6,97 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00074857
CAS nummer: 12036-10-1
EINECS: 234-840-6
UN: 1479
ADR: 5.1,II