Praseodymium(III) chloride heptahydrate 99,9%
Leverantör: Apollo Scientific
Varning
IN2965-250G IN2965-50G
APOSIN2965-250GEA 2575 SEK
APOSIN2965-250G APOSIN2965-50G
Praseodymium(III) chloride heptahydrate 99,9%
Praseodymium(III) chloride heptahydrate
Smältpunkt: 115 °C
Densitet: 2.25 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00149829
CAS nummer: 10025-90-8