Niob(V)oxid 99.99%
Leverantör: Apollo Scientific
Varning
Synonyms: Diniobium pentaoxide, Niobium pentoxide
IN2773-100G IN2773-500G
APOSIN2773-100GEA 2150 SEK
APOSIN2773-100G APOSIN2773-500G
Niob(V)oxid 99.99%
Niobium(V) oxide
Formel: Nb₂O₅
MW: 265,81 g/mol
Smältpunkt: 1485 °C
Densitet: 4,47 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011128
CAS nummer: 1313-96-8