Iron(II) trifluoromethanesulphonate
Leverantör: Apollo Scientific
Fara
Synonyms: Iron (II) Trifluoromethanesulphonate
IN20952-5G IN20952-25G
APOSIN20952-5GEA 550 SEK
APOSIN20952-5G APOSIN20952-25G
Iron(II) trifluoromethanesulphonate
Iron(II) trifluoromethanesulphonate
Formel: C₂F₆FeO₆S₂
MW: 353,99 g/mol
Smältpunkt: >300 °C
Storage Temperature: Omgivande
MDL Number: MFCD08460524
CAS nummer: 59163-91-6
UN: 3260
ADR: 8,II