Ammoniumjärn(II)sulfat hexahydrat ACS
Leverantör: Apollo Scientific
Varning
Synonyms: Ferrous Ammonium sulphate Hexahydrate, di-Ammonium iron (II) sulphate hexahydrate, Ferrous ammonium sulphate hexahydrate, Ammonium iron(Ⅱ) sulphate hexahydrate, Ammonium ferrous sulphate hexahydrate, Ammonium iron(II) sulphate hydrate
IN2044-250G IN2044-500G IN2044-2KG
APOSIN2044-250GEA 855 SEK
APOSIN2044-250G APOSIN2044-500G APOSIN2044-2KG
Ammoniumjärn(II)sulfat hexahydrat ACS
Ammoniumjärn(II)sulfat hexahydrat
Formel: (NH₄)₂Fe(SO₄)₂·6H₂O
MW: 392,14 g/mol
Smältpunkt: 100 °C (dec.)
Densitet: 1,864 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00150530
CAS nummer: 7783-85-9
EINECS: 233-151-8