Dysprosium trichloride hexahydrate 99,9%
Leverantör: Apollo Scientific
Varning
Synonyms: Dysprosium (III) chloride hexahydrate, Dysprosium(III) chloride hydrate
IN1639-100G IN1639-25G IN1639-250G
APOSIN1639-100GEA 7200 SEK
APOSIN1639-100G APOSIN1639-25G APOSIN1639-250G
Dysprosium trichloride hexahydrate 99,9%
Dysprosium trichloride hexahydrate
Formel: Cl₃Dy·6H₂O
MW: 376,95 g/mol
Smältpunkt: 718 °C
Storage Temperature: Omgivande
MDL Number: MFCD00010984
CAS nummer: 15059-52-6