Cesium hydroxide hydrate 99,9%

Leverantör: Apollo Scientific
Fara

Synonyms: Cesium hydroxide, hydrate

IN1450-100G IN1450-25G
APOSIN1450-100GEA 3100 SEK
APOSIN1450-100G APOSIN1450-25G
Cesium hydroxide hydrate 99,9%
Cesium hydroxide hydrate
Formel: HCsO·H₂O
MW: 167,93 g/mol
Smältpunkt: 272,3 °C
Densitet: 3,675 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00010964
CAS nummer: 12260-45-6
EINECS: 244-344-1
UN: 2682
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR