TES (2-(tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid) ≥99%
Leverantör: Apollo Scientific
Varning
Synonyms: TES Buffer, N-Tris(hydroxymethyl)methyl-2-aminoethanesulphonic acid
BIT3007-100G BIT3007-1KG BIT3007-500G
APOSBIT3007-100GEA 585 SEK
APOSBIT3007-100G APOSBIT3007-1KG APOSBIT3007-500G
TES (2-(tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid) ≥99%
TES (2-(tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid)
Biological buffer - useful range 6.8-8.2
  • Assay (by titration) >99%
  • Water (by Karl Fischer) <1.0%
Formel: C₆H₁₅NO₆S
MW: 229.25 g/mol
Smältpunkt: 225 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00007532
CAS nummer: 7365-44-8
EINECS: 230-906-3