TAPSO (2-hydroxy-3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid) 99
Leverantör: Apollo Scientific
Varning
Synonyms: TAPSO, 2-Hydroxy-3-{[2-hydroxy-1,1-bis(hydroxymethyl)-ethyl]amino}-1-propanesulphonic acid, 3-[N-Tris(hydroxymethyl)methylamino]-2-hydroxypropanesulphonic acid
BIT3003-100G BIT3003-1KG BIT3003-500G
APOSBIT3003-100GEA 615 SEK
APOSBIT3003-100G APOSBIT3003-1KG APOSBIT3003-500G
TAPSO (2-hydroxy-3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid) 99
TAPSO (2-hydroxy-3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid)
Biological buffer - useful range 7.0-8.2
  • Assay (by titration) >99%
  • Water (by Karl Fischer) <1.0%
U.S Patent No 4,169,950 (To Research Organics)
Formel: C₇H₁₇NO₇S
MW: 259.28 g/mol
Smältpunkt: 220…221 °C
Storage Temperature: Omgivande
MDL Number: MFCD00038352
CAS nummer: 68399-81-5