Tris-(hydroximetyl) aminometan hydroklorid 99,5
Leverantör: Apollo Scientific
Varning
Synonyms: Tris(hydroxymethyl)methylamine hydrochloride, 2-Amino-2-(hydroxymethyl)propane-1,3-diol hydrochloride, Trizma® hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, TRIS HCl, Tromethane hydrochloride, Tris-HCl, Tris(hydroxymethyl)methylammonium chloride, Trometamol hydrochloride, THAM HCl
BIT1513-1KG BIT1513-250G BIT1513-5KG
APOSBIT1513-1KGEA 1350 SEK
APOSBIT1513-1KG APOSBIT1513-250G APOSBIT1513-5KG
Tris-(hydroximetyl) aminometan hydroklorid 99,5
Tris-(hydroximetyl) aminometan hydroklorid
Biological buffer - useful range 7.7-9.1
  • Loss on drying: <0.5%
  • Heavy metals: <5ppm
  • pka @ 20°C:8.1
Formel: NH₂C(CH₂OH)₃·HCl
MW: 157.6 g/mol
Smältpunkt: 149…153 °C
Storage Temperature: Omgivande
MDL Number: MFCD00012590
CAS nummer: 1185-53-1
EINECS: 214-684-5
Merck Index: 12,09902