L (-)-Tyrosin ≥99.5%
Leverantör: Apollo Scientific
Varning
Synonyms: L-Tyrosine, (S)-(-)-Tyrosine, Tyrosine, H-Tyr-OH
BIT0721-100G BIT0721-1KG BIT0721-500G
APOSBIT0721-100GEA 295 SEK
APOSBIT0721-100G APOSBIT0721-1KG APOSBIT0721-500G
L (-)-Tyrosin ≥99.5%
L (-)-Tyrosin
Ph.Eur, pyrogen free
Formel: C₉H₁₁NO₃
MW: 181,19 g/mol
Smältpunkt: >300 °C (dec.)
Densitet: 1,46 g/cm³ (25 °C)
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002606
CAS nummer: 60-18-4
EINECS: 200-460-4