Tiaminhydroklorid ≥99%
Leverantör: Apollo Scientific
Varning
Synonyms: Aneurine hydrochloride, 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride hydrochloride, Thiaminium chloride, VitamineB1 hydrochloride
BIT0614-100G BIT0614-1KG BIT0614-250G
APOSBIT0614-100GEA 445 SEK
APOSBIT0614-100G APOSBIT0614-1KG APOSBIT0614-250G
Tiaminhydroklorid ≥99%
Tiaminhydroklorid
Formel: C₁₂H₁₈Cl₂N₄OS
MW: 337.27 g/mol
Smältpunkt: 247…249 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00012780
CAS nummer: 67-03-8
EINECS: 200-641-8
Merck Index: 13,09366