1,2-O-Isopropylidene-ɑ-D-glucofuranose
Leverantör: Apollo Scientific
Synonyms: (R)-1-((3aR,5R,6S,6aR)-6-Hydroxy-2,2-dimethyltetrahydrofuro[2,3-d][1,3]dioxol-5-yl)ethane-1,2-diol
BISY001-10G BISY001-25G BISY001-100G
APOSBISY001-10GEA 345 SEK
APOSBISY001-10G APOSBISY001-25G APOSBISY001-100G
1,2-O-Isopropylidene-ɑ-D-glucofuranose
1,2-O-Isopropylidene-alpha-D-glucofuranose
Formel: C₉H₁₆O₆
MW: 220,22 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00063244
CAS nummer: 18549-40-1