DB07268

Leverantör: Apollo Scientific

Synonyms: 2-[[2-(3-hydroxyanilino)pyrimidin-4-yl]amino]benzamide, 2-((2-((3-Hydroxyphenyl)amino)pyrimidin-4-yl)amino)benzamide

BISN0239-1MG BISN0239-10MG BISN0239-50MG
APOSBISN0239-1MGEA 1050 SEK
APOSBISN0239-1MG APOSBISN0239-10MG APOSBISN0239-50MG
DB07268
DB07268

MAPK/ERK Signalling Pathway

Formel: C₁₇H₁₅N₅O₂
MW: 321,34 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD26960941
CAS nummer: 929007-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR