LY-2584702 tosylate
Leverantör: Apollo Scientific
Synonyms:
4-(4-(4-(4-Fluoro-3-(trifluoromethyl)phenyl)-1-methyl-1H-imidazol-2-yl)piperidin-1-yl)-1H-pyrazolo[3,4-d]pyrimidine 4-methylbenzenesulphonate, LYS6K2, LY-2584702 tosylate salt
BISN0229-1MG
BISN0229-5MG
BISN0229-10MG
APOSBISN0229-1MGEA
855
SEK
APOSBISN0229-1MG
APOSBISN0229-5MG
APOSBISN0229-10MG
LY-2584702 tosylate
LY-2584702 tosylate
MAPK/ERK Signalling Pathway
Formel:
C₂₈H₂₇F₄N₇O₃S MW: 617,62 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD29054739 CAS nummer: 1082949-68-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...