TCS JNK 5a
Leverantör: Apollo Scientific
BISN0209-10MG BISN0209-50MG
APOSBISN0209-10MGEA 685 SEK
APOSBISN0209-10MG APOSBISN0209-50MG
TCS JNK 5a
TCS JNK 5a
MAPK/ERK Signalling Pathway
Formel: C₂₀H₁₆N₂OS
MDL Number: MFCD01005305
CAS nummer: 312917-14-9