Dabrafenib methanesulphonate salt

Leverantör: Apollo Scientific

Synonyms: Dabrafenib Mesylate, N-(3-(5-(2-Aminopyrimidin-4-yl)-2-(tert-butyl)thiazol-4-yl)-2-fluorophenyl)-2,6-difluorobenzenesulphonamide methanesulphonate, GSK-2118436 mesylate

BISN0172-1MG BISN0172-5MG BISN0172-10MG
APOSBISN0172-1MGEA 685 SEK
APOSBISN0172-1MG APOSBISN0172-5MG APOSBISN0172-10MG
Dabrafenib methanesulphonate salt
Dabrafenib methanesulphonate salt

MAPK/ERK Signalling Pathway

Formel: C₂₄H₂₄F₃N₅O₅S₃
MW: 615,67 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD20922872
CAS nummer: 1195768-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR