SGC0946

Leverantör: Apollo Scientific

Synonyms: 1-(3-((((2R,3S,4R,5R)-5-(4-Amino-5-bromo-7H-pyrrolo[2,3-d]pyrimidin-7-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl)(isopropyl)amino)propyl)-3-(4-(tert-butyl)phenyl)urea

BISN0137-1MG BISN0137-5MG BISN0137-10MG
APOSBISN0137-1MGEA 945 SEK
APOSBISN0137-1MG APOSBISN0137-5MG APOSBISN0137-10MG
SGC0946
SGC0946

SGC0946 is a selective inhibitor of DOT1L methyltransferase with IC50 of 03 nM and can kill mixed lineage leukaemia cells.

Formel: C₂₈H₄₀BrN₇O₄
MW: 618,577 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD26523136
CAS nummer: 1561178-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR