Verteporfin

Leverantör: Apollo Scientific

Synonyms: BPD-MA, 4R,4aS)-18-Ethenyl-4,4a-dihydro-3,4-bis(methoxycarbonyl)-4a,8,14,19-tetramethyl-24H,26H-benzo[b]porphine-9,13-dipropanoic acid monomethyl ester, (4R,4aS)-rel-18-Ethenyl-4,4a-dihydro-3,4-bis(methoxycarbonyl)-4a,8,14,19-tetramethyl-24H,26H-Benzo[b]porphine-9,13-dipropanoic acid monomethyl ester

BISN0133-1MG BISN0133-5MG BISN0133-10MG
APOSBISN0133-1MGEA 945 SEK
APOSBISN0133-1MG APOSBISN0133-5MG APOSBISN0133-10MG
Verteporfin
Verteporfin

Verteporfin a benzoporphyrin derivative monoacid ring A inhibits TEADYAP association and YAP-induced liver overgrowth It is also a potent second-generation photosensitizing agent.

Formel: C₄₁H₄₂N₄O₈
MW: 718,79 g/mol
Storage Temperature: Frys
MDL Number: MFCD28098202
CAS nummer: 129497-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR