MK-2206 dihydrochloride

Leverantör: Apollo Scientific

Synonyms: 8-(4-(1-Aminocyclobutyl)phenyl)-9-phenyl-[1,2,4]triazolo[3,4-f][1,6]naphthyridin-3(2H)-one dihydrochloride, MK-2206 2HCl

BISN0108-50MG BISN0108-100MG
APOSBISN0108-50MGEA 3250 SEK
APOSBISN0108-50MG APOSBISN0108-100MG
MK-2206 dihydrochloride
MK-2206 dihydrochloride

MK-2206 2HCl is a highly selective inhibitor of Akt1/2/3 with IC50 of 8 nM/12 nM/65 nM, respectively, showing no inhibitory activities against 250 other protein kinases observed.

Formel: C₂₅H₂₁N₅O·2HCl
MW: 480,4 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD14584463
CAS nummer: 1032350-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR