Radicicol

Leverantör: Apollo Scientific
Fara

BIR0140-10MG BIR0140-1MG BIR0140-5MG
APOSBIR0140-10MGEA 7700 SEK
APOSBIR0140-10MG APOSBIR0140-1MG APOSBIR0140-5MG
Radicicol
Radicicol
Formel: C₁₈H₁₇ClO₆
MW: 364.8 g/mol
Storage Temperature: Frys
CAS nummer: 12772-57-5
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR