(-)-Quinine hydrochloride dihydrate
Leverantör: Apollo Scientific
Fara
Synonyms: Quinine hydrochloride dihydrate, Quinine monohydrochloride dihydrate, (R)-(6-Methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol hydrochloride dihydrate
BIQ1204-100G BIQ1204-10G BIQ1204-25G
APOSBIQ1204-100GEA 1550 SEK
APOSBIQ1204-100G APOSBIQ1204-10G APOSBIQ1204-25G
(-)-Quinine hydrochloride dihydrate
(-)-Quinine hydrochloride dihydrate
Formel: C₂₀H₂₅ClN₂O₂·2H₂O
MW: 396,91 g/mol
Smältpunkt: 115…116 °C (dec.)
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00151248
CAS nummer: 6119-47-7
EINECS: 205-001-1