Mal-amido-PEG6-acid
Leverantör: Apollo Scientific
Synonyms: 1-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)-3-oxo-7,10,13,16,19,22-hexaoxa-4-azapentacosan-25-oic acid
BIPG1474-1G BIPG1474-250MG
APOSBIPG1474-1GEA 4475 SEK
APOSBIPG1474-1G APOSBIPG1474-250MG
Mal-amido-PEG6-acid
Mal-amido-PEG6-acid
Formel: C₂₂H₃₆N₂O₁₁
MW: 504.53 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD21363276
CAS nummer: 1334177-79-5