14-Azido-3,6,9,12-tetraoxatetradecanol
Leverantör: Apollo Scientific
Synonyms: PEG5-Azide
BIPG1118-250MG BIPG1118-1G
APOSBIPG1118-250MGEA 2125 SEK
APOSBIPG1118-250MG APOSBIPG1118-1G
14-Azido-3,6,9,12-tetraoxatetradecanol
14-Azido-3,6,9,12-tetraoxatetradecanol
Formel: C₁₀H₂₁N₃O₅
MW: 263,29 g/mol
Flampunkt: 100 °C
Storage Temperature: Kylskåp
MDL Number: MFCD20134131
CAS nummer: 86770-68-5