Amino-PEG4-t-butyl ester

Leverantör: Apollo Scientific
Varning

BIPG1073-500MG BIPG1073-250MG BIPG1073-1G
APOSBIPG1073-500MGEA 2850 SEK
APOSBIPG1073-500MG APOSBIPG1073-250MG APOSBIPG1073-1G
Amino-PEG4-t-butyl ester
Amino-PEG4-t-butyl ester
Formel: C₁₅H₃₁NO₆
MW: 321,41 g/mol
Storage Temperature: Omgivande
CAS nummer: 581065-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR