Amino-PEG4-acid

Leverantör: Apollo Scientific
Varning

Synonyms: NH2-PEG4-CH2CH2COOH, (PEO)4-Aminopropionic acid, 1-Amino-3,6,9,12-tetraoxapentadecan-15-oic acid, Tetraethylene glycol α-amino-ω-propionic acid

BIPG1052-100MG BIPG1052-250MG BIPG1052-500MG
APOSBIPG1052-100MGEA 1050 SEK
APOSBIPG1052-100MG APOSBIPG1052-250MG APOSBIPG1052-500MG
Amino-PEG4-acid
Amino-PEG4-acid
Formel: C₁₁H₂₃NO₆
MW: 265,30 g/mol
Storage Temperature: Frys
MDL Number: MFCD11041129
CAS nummer: 663921-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR